missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERO1LB Antibody (M37-P5D11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50389
This item is not returnable.
View return policy
Description
ERO1LB Monoclonal antibody specifically detects ERO1LB in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| ERO1LB | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin | |
| DKFZp779C1042, DKFZp779I0141, EC 1.8.4, EC 1.8.4.-, Endoplasmic oxidoreductin-1-like protein B, ERO1-L(beta), ERO1-L-beta, ERO1-like beta (S. cerevisiae), ERO1-like protein beta, FLJ11003, oxidoreductin-1-L-beta | |
| Peptide - LAPSRGEDDG | |
| 0.1 mL | |
| Signal Transduction | |
| 56605 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| M37-P5D11 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction