missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ERP27 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558868
This item is not returnable.
View return policy
Description
Immunogen sequence: ILVDSGMKEN GKVISFFKLK ESQLPALAIY QTLDDEWDTL PTAEVSVEHV QNFCDGFLSG KLLKENRESE GKTPKV Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 57%.
Specifications
| ERP27 | |
| Polyclonal | |
| Unconjugated | |
| ERP27 | |
| C12orf46; endoplasmic reticulum protein 27; endoplasmic reticulum protein 27 kDa; endoplasmic reticulum resident protein 27; ER protein 27; ERP27; Inactive protein disulfide-isomerase 27; PDIA8; protein disulfide isomerase family A, member 8; UNQ781/PRO1575 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 121506 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96DN0 | |
| ERP27 | |
| Recombinant protein corresponding to Human ERP27. Recombinant protein control fragment (Product #RP-97752). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction