missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERP29 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99200-100ul
This item is not returnable.
View return policy
Description
ERP29 Polyclonal antibody specifically detects ERP29 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| ERP29 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:500-1:2000 | |
| C12orf8chromosome 12 open reading frame 8, endoplasmic reticulum lumenal protein ERp28, endoplasmic reticulum protein 29, Endoplasmic reticulum resident protein 28, endoplasmic reticulum resident protein 29, ERp28Endoplasmic reticulum resident protein 31, ERp29, ERp31ERP28, PDIA9, PDI-DB, protein disulfide isomerase family A, member 9 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human ERP29. | |
| 100 μg | |
| Core ESC Like Genes, Membrane Trafficking and Chaperones, Stem Cell Markers | |
| 10961 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction