missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERP29 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67675-1-IG-20UL
This item is not returnable.
View return policy
Description
The ERP29 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ERP29. This antibody recognizes human, mouse, rat antigen. The ERP29 antibody has been validated for the following applications: WB, IHC, IF-P, ELISA analysis.Specifications
| ERP29 | |
| Monoclonal | |
| 1100 μg/mL | |
| C12orf8; ERp28; ERp31; PDI-DB; PDIA9 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 29 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 1H8B2 | |
| Unconjugated | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 10961 | |
| Store at -20°C. | |
| Monoclonal Antibody | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction