Learn More
ERP44 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-97000V
Code produit. 18067276
  • € 415.00 / 0.10mL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
ERP44 Polyclonal specifically detects ERP44 in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY
Spécification
Spécification
ERP44
Polyclonal
Western Blot, ELISA
endoplasmic reticulum protein 44, endoplasmic reticulum resident protein 44 kDa, ER protein 44, ERp44, KIAA0573endoplasmic reticulum resident protein 44, PDIA10, protein disulfide isomerase family A, member 10, thioredoxin domain containing 4 (endoplasmic reticulum), Thioredoxin domain-containing protein 4, TXNDC4
Produced in rabbits immunized with purified, recombinant ERP44 (NP_055866.1; Met1-Asp402).
0.1 mL
Primary
Human
Purified
Western Blot, ELISA
DyLight 405
50mM Sodium Borate
Rabbit
Affinity purified
RUO
23071
Store at 4°C in the dark.