missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERP44 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97000IR
This item is not returnable.
View return policy
Description
ERP44 Polyclonal specifically detects ERP44 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| ERP44 | |
| Polyclonal | |
| Western Blot, ELISA | |
| endoplasmic reticulum protein 44, endoplasmic reticulum resident protein 44 kDa, ER protein 44, ERp44, KIAA0573endoplasmic reticulum resident protein 44, PDIA10, protein disulfide isomerase family A, member 10, thioredoxin domain containing 4 (endoplasmic reticulum), Thioredoxin domain-containing protein 4, TXNDC4 | |
| Produced in rabbits immunized with purified, recombinant ERP44 (NP_055866.1; Met1-Asp402). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 23071 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction