Learn More
Invitrogen™ ERP44 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109693
Description
Immunogen sequence: WCRFSQMLHP IFEEASDVIK EEFPNENQVV FARVDCDQHS DIAQRYRISK YPTLKLFRNG MMMKREYRGQ RSVKALADYI RQQKSDPIQE IRDLAEITTL DRSKRNIIGY FEQKDSDNYR VFERVANILH DDCAFLSAFG DVSKPERYSG DNIIYKPPGH SAPDMVYLGA MTNFDVTYNW IQDKCVPLVR EITFENGEEL TEEGLPFLIL FHMKEDTESL EIFQNEVARQ LISEKGTINF LHADCDKFRH PLLHIQKTPA DCPVIAIDSF RHMYVFGDFK DVLIPGKLKQ FVFDLHSGKL HREFHHGPDP TDTAPGEQAQ DVASSPPESS FQKLAPSEYR YTLLRDRDEL.
Mediates thiol-dependent retention in the early secretory pathway, forming mixed disulfides with substrate proteins through its conserved CRFS motif. Inhibits the calcium channel activity of ITPR1. May have a role in the control of oxidative protein folding in the endoplasmic reticulum. Required to retain ERO1L and ERO1LB in the endoplasmic reticulum.
Specifications
| ERP44 | |
| Polyclonal | |
| Unconjugated | |
| ERP44 | |
| 1110001E24Rik; AI849526; AL033348; BWK4; endoplasmic reticulum protein 44; endoplasmic reticulum resident protein 44; endoplasmic reticulum resident protein 44 kDa; endoplasmic reticulum resident protein 44kDa; endoplasmic reticulum resident protein ERp44; ER protein 44; Erp44; KIAA0573; OTTHUMP00000063799; PDIA10; protein disulfide isomerase family A, member 10; thioredoxin domain containing 4 (endoplasmic reticulum); thioredoxin domain-containing protein 4; Txndc4; UNQ532/PRO1075 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 23071, 298066, 76299 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.71 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9BS26, Q9D1Q6 | |
| ERP44 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 57-406 of human ERP44 (NP_055866.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.