missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ERp57 Monoclonal Antibody (Map.ERp57), APC

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA545227
This item is not returnable.
View return policy
Description
0.5 μg/mL of MA5-45227 was sufficient for detection of ERp57 in 10 μg of heat shock Heal Lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 57kDa.
This gene encodes a protein of the endoplasmic reticulum that interacts with lectin chaperones calreticulin and calnexin to modulate folding of newly synthesized glycoproteins. The protein was once thought to be a phospholipase; however, it has been demonstrated that the protein actually has protein disulfide isomerase activity. It is thought that complexes of lectins and this protein mediate protein folding by promoting formation of disulfide bonds in their glycoprotein substrates.
Specifications
| ERp57 | |
| Monoclonal | |
| 1 mg/mL | |
| 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4 | |
| P11598, P27773, P30101, P38657 | |
| PDIA3 | |
| Human recombinant ERp57 (Grp58). | |
| 200 μg | |
| Primary | |
| Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Hamster, Pig, Guinea Pig | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| Map.ERp57 | |
| APC | |
| PDIA3 | |
| 58 kDa glucose-regulated protein; 58 kDa microsomal protein; 58kDa; Disulfide isomerase ER-60; endoplasmic reticulum P58; endoplasmic reticulum protein; endoplasmic reticulum resident protein 57; Endoplasmic reticulum resident protein 60; epididymis secretory protein Li 269; epididymis secretory sperm binding protein Li 93n; ER p57; ER protein 57; ER protein 60; ER60; ER-60 protease; Erp; ERp 57; ERp57; ERP57 protein; ERP60; ERp61; glucose regulated protein 58; glucose regulated protein 58kD; glucose regulated protein, 58 kDa; glucose regulated protein, 58kDa; GRP 57; GRP 58; GRP57; Grp58; grp-58; HEL-S-269; HEL-S-93n; HIP-70; HsT17083; I79_003765; oxidoreductase ERp57; p58; PDI; PDIA 3; PDIA3; PDI-Q2; phospholipase C, alpha; phospholipase C-alpha; PI PLC; PI-PLC; PLC alpha; PLC[a]; Plca; protein disulfide isomerase associated 3; protein disulfide isomerase family A member 3; protein disulfide isomerase family A, member 3; protein disulfide isomerase-associated 3; protein disulfide-isomerase A3; Q-2 | |
| Mouse | |
| Protein G | |
| RUO | |
| 100156204, 100328685, 100689343, 100725599, 14827, 281803, 2923, 29468, 478279 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction