missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERp57/PDIA3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-84797
This item is not returnable.
View return policy
Description
ERp57/PDIA3 Polyclonal specifically detects ERp57/PDIA3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
Specifications
| ERp57/PDIA3 | |
| Polyclonal | |
| Western Blot 0.04 - 0.4 μg/mL, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000, Knockdown Validated | |
| P30101 | |
| PDIA3 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:HMTEDNKDLIQGKDLLIAYYDVDYEKNAKGSNYWRNRVMMVAKKFLDAGHKLNFAVASRKTFSHELSDFGLESTAGEIPVVAIRTAKGEKFVMQEEFSRDGKALERFLQDYFDGNLKRYLKSEPIPESNDGP | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| Disulfide isomerase ER-60, EC 5.3.4.1, endoplasmic reticulum P58, Endoplasmic reticulum resident protein 57, Endoplasmic reticulum resident protein 60, ER protein 60, ER60, ERP57, ERp5758 kDa glucose-regulated protein, ERP60, ERp6058 kDa microsomal protein, ERp61, glucose regulated protein, 58kDa, GRP57, GRP58ER protein 57, HsT17083, P58, phospholipase C-alpha, PI-PLC, protein disulfide isomerase family A, member 3, protein disulfide isomerase-associated 3, protein disulfide-isomerase A3 | |
| Rabbit | |
| 57 kDa | |
| 0.1 mL | |
| Cellular Markers, ER Markers, Membrane Trafficking and Chaperones | |
| 2923 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction