missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERp57/PDIA3 Antibody (OTI3E1), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70848AF700
This item is not returnable.
View return policy
Description
ERp57/PDIA3 Monoclonal specifically detects ERp57/PDIA3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| ERp57/PDIA3 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PDIA3 | |
| Recombinant protein fragment corresponding to amino acids PDIA3(NP_005304) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| OTI3E1 | |
| Western Blot, Immunohistochemistry | |
| Disulfide isomerase ER-60, EC 5.3.4.1, endoplasmic reticulum P58, Endoplasmic reticulum resident protein 57, Endoplasmic reticulum resident protein 60, ER protein 60, ER60, ERP57, ERp5758 kDa glucose-regulated protein, ERP60, ERp6058 kDa microsomal protein, ERp61, glucose regulated protein, 58kDa, GRP57, GRP58ER protein 57, HsT17083, P58, phospholipase C-alpha, PI-PLC, protein disulfide isomerase family A, member 3, protein disulfide isomerase-associated 3, protein disulfide-isomerase A3 | |
| Mouse | |
| Affinity Purified | |
| Cellular Markers, ER Markers, Membrane Trafficking and Chaperones | |
| 2923.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction