missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERp72 Antibody (004), Alexa Fluor™ 488, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90207AF488
This item is not returnable.
View return policy
Description
ERp72 Monoclonal specifically detects ERp72 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| ERp72 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunofluorescence | |
| 4 | |
| Immunocytochemistry/Immunofluorescence, Immunofluorescence | |
| Endoplasmic reticulum resident protein 70, Endoplasmic reticulum resident protein 72, ER protein 70, ERp70, ERP70ER protein 72, ERp72, ERp-72, ERP72EC 5.3.4.1, intestinal-related), protein disulfide isomerase family A, member 4, protein disulfide isomerase related protein (calcium-binding protein, protein disulfide isomerase-associated 4, protein disulfide-isomerase A4 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human ERp72 (P13667; Met1-Thr641). | |
| 0.1 mL | |
| Core ESC Like Genes, Membrane Trafficking and Chaperones, Stem Cell Markers | |
| 9601 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction