missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERR beta/NR3B2 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99541AF405
This item is not returnable.
View return policy
Description
ERR beta/NR3B2 Polyclonal antibody specifically detects ERR beta/NR3B2 in Mouse samples. It is validated for Western Blot
Specifications
| ERR beta/NR3B2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| Alexa Fluor 405 | |
| estrogen-related receptor beta | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ERR beta/NR3B2 | |
| 0.1 mL | |
| Cancer | |
| 2103 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction