missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERR gamma/NR3B3 Antibody (OTI1E5), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70691CL1
This item is not returnable.
View return policy
Description
ERR gamma/NR3B3 Monoclonal antibody specifically detects ERR gamma/NR3B3 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| ERR gamma/NR3B3 | |
| Monoclonal | |
| CoraFluor 1 | |
| DKFZp781L1617, ERR gamma-2, ERR3, ERRG2, ERRgamma, Estrogen receptor-related protein 3, estrogen-related receptor gamma, FLJ16023, KIAA0832, NR3B3, Nuclear receptor subfamily 3 group B member 3 | |
| Human recombinant protein fragment corresponding to amino acids 24-263 of human ESRRG(NP_001429) produced in E.coli. | |
| 0.1 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 2104 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Immunohistochemistry, Western Blot | |
| OTI1E5 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction