missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERRF Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81715AF488
This item is not returnable.
View return policy
Description
ERRF Polyclonal antibody specifically detects ERRF in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| ERRF | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 488 | |
| chromosome 1 open reading frame 64, ERRF, MGC24047, putative uncharacterized protein C1orf64, RP11-5P18.4 | |
| ERRF antibody was raised against a 15 amino acid peptide near the carboxy terminus of human ERRF . The immunogen is located within the last 50 amino acids of ERRF. Amino Acid Squence: CPQEVPEAKGKPVKA | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 149563 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction