missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ertapenem sodium, 85%

Catalog Number p-7077849
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
153773-82-1
C22H24N3NaO7S
497.5
ZXNAQFZBWUNWJM-HRXMHBOMSA-M
1-azabicyclo 3.2.0 hept-2-ene-2-carboxylic acid, 3-3s,5s-5-3-ca rboxyphenyl amino carbonyl-3-pyrrolidinyl thio-6-1r-1-hydroxyethy l-4-methyl-7-oxo-, monosodium salt, 4r,5s,6s
131675376
CHEBI:60070
sodium;(2S,4S)-4-[[(4R,5S,6S)-2-carboxy-6-[(1R)-1-hydroxyethyl]-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-en-3-yl]sulfanyl]-N-(3-carboxyphenyl)pyrrolidine-2-carboximidate
CC1C2C(C(=O)N2C(=C1SC3CC(NC3)C(=NC4=CC=CC(=C4)C(=O)O)[O-])C(=O)O)C(C)O.[Na+]
This item is not returnable. View return policy
153773-82-1
C22H24N3NaO7S
497.5
ZXNAQFZBWUNWJM-HRXMHBOMSA-M
1-azabicyclo 3.2.0 hept-2-ene-2-carboxylic acid, 3-3s,5s-5-3-ca rboxyphenyl amino carbonyl-3-pyrrolidinyl thio-6-1r-1-hydroxyethy l-4-methyl-7-oxo-, monosodium salt, 4r,5s,6s
131675376
CHEBI:60070
sodium;(2S,4S)-4-[[(4R,5S,6S)-2-carboxy-6-[(1R)-1-hydroxyethyl]-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-en-3-yl]sulfanyl]-N-(3-carboxyphenyl)pyrrolidine-2-carboximidate
CC1C2C(C(=O)N2C(=C1SC3CC(NC3)C(=NC4=CC=CC(=C4)C(=O)O)[O-])C(=O)O)C(C)O.[Na+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 230.0°C to 234.0°C
Loss on Drying 15% to 19%
Infrared Spectrum Authentic
Assay Percent Range 80% min. (HPLC)
Quantity 250 mg
Beilstein 15,3731
Formula Weight 497.5
Percent Purity 85%
Chemical Name or Material Ertapenem sodium
compliance-icons
Product Identifier
  • Ertapenem sodium
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'