missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Erythropoietin R Antibody (38409) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB307JF669
This item is not returnable.
View return policy
Description
Erythropoietin R Monoclonal antibody specifically detects Erythropoietin R in Human samples. It is validated for Flow Cytometry,Immunoprecipitation,Western Blot
Specifications
| Erythropoietin R | |
| Monoclonal | |
| Janelia Fluor 669 | |
| EPO-R, erythropoietin receptor, MGC138358 | |
| Mouse myeloma cell line NS0-derived recombinant human Erythropoietin R, Pro26-Pro250, Accession # P19235 | |
| 0.1 mL | |
| Angiogenesis, Cancer, Immunology, Phospho Specific, Signal Transduction | |
| 2057 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, Immunoprecipitation, Western Blot | |
| 38409 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction