missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ESAM Antibody (1021723), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB42042C
This item is not returnable.
View return policy
Description
ESAM Monoclonal antibody specifically detects ESAM in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| ESAM | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 90952 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, CyTOF | |
| 1021723 | |
| Flow Cytometry, CyTOF-ready | |
| 2310008D05Rik, endothelial cell adhesion molecule, endothelial cell-selective adhesion molecule, HUEL (C4orf1)-interacting protein, LP4791 protein, W117m | |
| Mouse myeloma cell line NS0-derived recombinant human ESAM, Gln30-Ala247, Accession # Q96AP7 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction