Learn More
Invitrogen™ Escherichia Virus Capsid Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144462
This item is not returnable.
View return policy
Description
Escherichia Virus Capsid Polyclonal Antibody for Western Blot, ELISA
Capsid protein self-assembles to form an icosahedral capsid with a T=3 symmetry, about 26 nm in diameter, and consisting of 89 capsid proteins dimers (178 capsid proteins) (PubMed:27671640, PubMed:19913556). Involved in viral genome encapsidation through the interaction between a capsid protein dimer and the multiple packaging signals present in the RNA genome (PubMed:8943226, PubMed:27671640). Binding of the capsid proteins to the viral RNA induces a conformational change required for efficient T=3 shell formation (PubMed:19913556). The capsid contains also 1 copy of the A2 maturation protein (PubMed:27671640).
Specifications
| Escherichia Virus Capsid | |
| Polyclonal | |
| Unconjugated | |
| Capsid protein | |
| Recombinant Escherichia phage Qbeta (Bacteriophage Q-beta) CAPSD protein. | |
| 100 μL | |
| Primary | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 2.65 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| Rabbit | |
| Protein A/G | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG |