Learn More
Invitrogen™ Escherichia Virus Capsid Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5144462
Product Code. 17910663
  • € 371.00 / 100µL
Description
Escherichia Virus Capsid Polyclonal Antibody for Western Blot, ELISA
Capsid protein self-assembles to form an icosahedral capsid with a T=3 symmetry, about 26 nm in diameter, and consisting of 89 capsid proteins dimers (178 capsid proteins) (PubMed:27671640, PubMed:19913556). Involved in viral genome encapsidation through the interaction between a capsid protein dimer and the multiple packaging signals present in the RNA genome (PubMed:8943226, PubMed:27671640). Binding of the capsid proteins to the viral RNA induces a conformational change required for efficient T=3 shell formation (PubMed:19913556). The capsid contains also 1 copy of the A2 maturation protein (PubMed:27671640).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Escherichia Virus Capsid
Polyclonal
Unconjugated
Capsid protein
Recombinant Escherichia phage Qbeta (Bacteriophage Q-beta) CAPSD protein.
100 μL
Primary
-20°C or -80°C if preferred
Liquid
ELISA, Western Blot
2.65 mg/mL
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Rabbit
Protein A/G
RUO
Bacteria