missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ESF1 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-71920PCP
This item is not returnable.
View return policy
Description
ESF1 Polyclonal antibody specifically detects ESF1 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| ESF1 | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| ABT1-associated protein, ABTAP, bA526K24.1, C20orf6, chromosome 20 open reading frame 6, ESF1 homolog, ESF1, nucleolar pre-rRNA processing protein, homolog (S. cerevisiae), FLJ20368 | |
| The immunogen for this product maps to a region between residue 801 and 851 of human ABT1-Associated Protein using the numbering given in entry NP_057733.2 (GeneID 51575). | |
| 0.1 mL | |
| Stem Cell Markers | |
| 51575 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| PerCP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction