missing translation for 'onlineSavingsMsg'
Learn More

ESI-09, MedChemExpress

Product Code. 16479677
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16479677 5 mg 5mg
16437148 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16479677 Supplier Medchem Express Supplier No. HY16704

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ESI-09 is a novel noncyclic nucleotide EPAC antagonist with IC50 values of 3.2 and 1.4 μM for EPAC1 and EPAC2, respectively.

ESI-09 is a novel noncyclic nucleotide EPAC antagonist with IC50 values of 3.2 and 1.4 μM for EPAC1 and EPAC2, respectively.

Specifications

Chemical Name or Material ESI-09
Physical Form Powder
CAS 263707-16-0
Color Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C16H15ClN4O2
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : ≥ 47 mg/mL (142.09 mM)
SMILES O=C(C1=NOC(C(C)(C)C)=C1)/C(C#N)=N/NC2=CC(Cl)=CC=C2
Molecular Weight (g/mol) 330.77
Formula Weight 330.77
Percent Purity 98.07%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.