missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNFAIP8 Mouse anti-Human, Clone: OTI1H9, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810682S
This item is not returnable.
View return policy
Description
TNFAIP8 Monoclonal antibody specifically detects TNFAIP8 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| TNFAIP8 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O95379 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 25816 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1H9 | |
| Unconjugated | |
| Tnfaip8 | |
| AA987150; E130304C20Rik; ENSMUSG00000073567; GG2-1; Gm10539; head and neck tumor and metastasis-related protein; MDC3.13; MDC-3.13; Nded; NF-kappa-B-inducible DED-containing protein; SCC S2; SCCS2; SCC-S2; Ssc-2; TNF alpha induced protein 8; TNF alpha-induced protein 8; Tnfaip8; TNFalpha IP8; TNFalpha-IP 8; TNFalphaIP8; TNF-induced protein; TNF-induced protein GG2-1; Tumor necrosis factor alpha-induced protein 8; tumor necrosis factor, alpha induced protein 8; tumor necrosis factor, alpha-induced protein 8 | |
| Full length human recombinant protein of human TNFAIP8 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction