Learn More
ESR2 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 30694-1-AP-20UL
 View more versions of this product
Product Code. 18055495
  • € 145.00 / 20µL
Description
The ESR2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ESR2. This antibody recognizes human antigen. The ESR2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
ESR2
800 μg/mL
Rabbit
Primary
Human
150 kDa
IgG
Western Blot, ELISA
ER BETA, Erb, ESR BETA, ESR2, ESRB, ESTRB, estrogen receptor 2 (ER beta), Estrogen receptor beta, NR3A2
20 μL
2100
Store at -20°C. Stable for one year after shipment.