Learn More
ESR2 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30694-1-AP-20UL
This item is not returnable.
View return policy
Description
The ESR2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ESR2. This antibody recognizes human antigen. The ESR2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| ESR2 | |
| 800 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| ER BETA, Erb, ESR BETA, ESR2, ESRB, ESTRB, estrogen receptor 2 (ER beta), Estrogen receptor beta, NR3A2 | |
| 20 μL | |
| 2100 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |