missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ESRP2 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76537AF532
This item is not returnable.
View return policy
Description
ESRP2 Polyclonal antibody specifically detects ESRP2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| ESRP2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 80004 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 532 | |
| epithelial splicing regulatory protein 2, FLJ21918, FLJ22248, RBM35BRNA-binding protein 35B, RNA binding motif protein 35A, RNA binding motif protein 35B, RNA-binding motif protein 35B | |
| Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human RBM35B. The immunogen is located within the last 50 amino acids of RBM35B. Amino Acid Squence: PADDYTSLMPVGDPPRT | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction