missing translation for 'onlineSavingsMsg'
Learn More

VSNL1 Mouse anti-Human, Clone: UMAB116, liquid, UltraMAB™

Mouse Monoclonal Antibody

Brand:  Origene Technologies UM870035

Encompass_Preferred

Product Code. 17590105

  • € 271.04 / 30µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This gene is a member of the visinin/recoverin subfamily of neuronal calcium sensor proteins. The encoded protein is strongly expressed in granule cells of the cerebellum where it associates with membranes in a calcium-dependent manner and modulates intracellular signaling pathways of the central nervous system by directly or indirectly regulating the activity of adenylyl cyclase. Alternatively spliced transcript variants have been observed, but their full-length nature has not been determined.
Specifications

Specifications

VSNL1
Monoclonal
0.5-1.0 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P62760
VSNL1
Human recombit protein fragment corresponding to amino acids 2-191 of human VSNL1 produced in E.coli.
30 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin)
UMAB116
Unconjugated
VSNL1
HLP3, HPCAL3, HUVISL1, VILIP, VILIP-1
Mouse
Affinity Chromatography
RUO
7447
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.