missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ESRRA Monoclonal Antibody (OTI2C12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808637
This item is not returnable.
View return policy
Description
Estrogen-related receptor alpha (ERR alpha), a NR3 Steroid Receptor, was isolated based on sequence similarity in its DNA-binding domain to estrogen receptor alpha (ER alpha). ERR alpha has been shown to regulate the promoters of lactoferrin, medium-chain acyl CoA dehydrogenase, osteopontin, and thyroid receptor alpha, and it may affect cellular energy balance and bone formation.Specifications
| ESRRA | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P11474 | |
| ESRRA | |
| Human recombit protein fragment corresponding to amino acids 1-205 of human ESRRA produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2C12 | |
| Unconjugated | |
| ESRRA | |
| ERR1, ERRa, ERRalpha, ESRL1, NR3B1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 2101 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction