Learn More
ESRRB Monoclonal Antibody (OTI12H2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA805749
Encompass_Preferred
 View more versions of this product
Product Code. 17535743
  • POA / 100µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
ESRRB (steroid hormone receptor ERR2) plays a role in maintaice of self-renewal and pluripotency of embryonic and trophoblast stem cells through different signaling pathways including FGF signaling pathway and Wnt signaling pathways. Upon FGF signaling pathway activation ESRRB interacts with KDM1A by directly binding to enhancer sites of ELF5 and EOMES and activates their transcription leading to self-renewal of trophoblast stem cells. ESSRB also regulates expression of multiple rod-specific genes and is required for survial of this cell type. Defects in the gene can cause deafness, autosomal recessive, 35 (DFNB35).
Specifications
Specifications
ESRRB
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O95718
ESRRB
Human recombit protein fragment corresponding to amino acids 1-252 of human ESRRB produced in E.coli.
100 μL
Primary
Human
Antibody
IgG2b
Western Blot
OTI12H2
Unconjugated
ESRRB
DFNB35, ERR2, ERRb, ESRL2, NR3B2
Mouse
Affinity Chromatography
RUO
2103
-20° C, Avoid Freeze/Thaw Cycles