missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ESRRB Monoclonal Antibody (OTI12H2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805749S
This item is not returnable.
View return policy
Description
ESRRB Monoclonal antibody specifically detects ESRRB in Human samples. It is validated for Western BlotSpecifications
| ESRRB | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P05019 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 3479 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI12H2 | |
| Unconjugated | |
| IGF1 | |
| DFNB35; ERR beta-2; ERR2; Err-2; ERRb; ERRB2; ERRbeta; ERR-beta; ERRbeta-2; ESRL2; ESRRB; estrogen receptor related 2; estrogen receptor-like 2; estrogen related receptor beta; estrogen related receptor, beta; estrogen-related nuclear receptor beta; estrogen-related receptor beta; estrogen-related receptor, beta; Estrrb; NR3B2; nuclear receptor ERRB2; nuclear receptor subfamily 3 group B member 2; orphan nuclear receptor; steroid hormone receptor ERR2 | |
| Human recombinant protein fragment corresponding to amino acids 1-151 of human IGF1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction