missing translation for 'onlineSavingsMsg'
Learn More

ESRRB Monoclonal Antibody (OTI1E6), TrueMAB™, OriGene

Product Code. 17575393
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17575393 30 μL 30µL
17555393 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17575393 Supplier Origene Technologies Supplier No. TA805455S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ESRRB Monoclonal antibody specifically detects ESRRB in Human samples. It is validated for Western Blot

Specifications

Antigen ESRRB
Applications Western Blot
Classification Monoclonal
Clone OTI1E6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ESRRB
Gene Accession No. O95718
Gene Alias DFNB35; ERR beta-2; ERR2; Err-2; ERRb; ERRB2; ERRbeta; ERR-beta; ERRbeta-2; ESRL2; ESRRB; estrogen receptor related 2; estrogen receptor-like 2; estrogen related receptor beta; estrogen related receptor, beta; estrogen-related nuclear receptor beta; estrogen-related receptor beta; estrogen-related receptor, beta; Estrrb; NR3B2; nuclear receptor ERRB2; nuclear receptor subfamily 3 group B member 2; orphan nuclear receptor; steroid hormone receptor ERR2
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-252 of human ESRRB produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2103
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.