missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ESRRB Monoclonal Antibody (OTI2F11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805454
This item is not returnable.
View return policy
Description
ESRRB (steroid hormone receptor ERR2) plays a role in maintaice of self-renewal and pluripotency of embryonic and trophoblast stem cells through different signaling pathways including FGF signaling pathway and Wnt signaling pathways. Upon FGF signaling pathway activation ESRRB interacts with KDM1A by directly binding to enhancer sites of ELF5 and EOMES and activates their transcription leading to self-renewal of trophoblast stem cells. ESSRB also regulates expression of multiple rod-specific genes and is required for survial of this cell type. Defects in the gene can cause deafness, autosomal recessive, 35 (DFNB35).Specifications
| ESRRB | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O95718 | |
| ESRRB | |
| Human recombit protein fragment corresponding to amino acids 1-252 of human ESRRB produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| OTI2F11 | |
| Unconjugated | |
| ESRRB | |
| DFNB35, ERR2, ERRb, ESRL2, NR3B2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 2103 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction