missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ESRRG Monoclonal Antibody (OTI1D10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504963
This item is not returnable.
View return policy
Description
Estrogen-related receptor gamma (ERRG, ERR3, NR3B3) is an orphan member of the nuclear hormone receptor superfamily. ERRG acts as a transcription activator in the absence of a bound ligand. It binds specifically to an estrogen response element and activates reporter genes controlled by estrogen response elements. ERRG also induces the expression of PERM1 in the skeletal muscle.Specifications
| ESRRG | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P62508 | |
| ESRRG | |
| Human recombit protein fragment corresponding to amino acids 24-263 of human ESRRG produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI1D10 | |
| Unconjugated | |
| ESRRG | |
| ERR3, ERRgamma, NR3B3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 2104 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction