Learn More
Invitrogen™ ESRRG Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595510
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human PANC-1 whole cell, human 22RV1 whole cell, human COLO-320 whole cell, human SW620 whole cell, mouse SP20 whole cell. IHC: mouse brain tissue, rat cardiac muscle tissue. ICC/IF: A549 cell. Flow: U20S cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Estrogen-related receptor gamma (ERRG, ERR3, NR3B3) is an orphan member of the nuclear hormone receptor superfamily. ERRG acts as a transcription activator in the absence of a bound ligand. It binds specifically to an estrogen response element and activates reporter genes controlled by estrogen response elements. ERRG also induces the expression of PERM1 in the skeletal muscle.
Specifications
| ESRRG | |
| Polyclonal | |
| Unconjugated | |
| Esrrg | |
| DKFZp781L1617; ERR gamma; ERR gamma-2; ERR3; errg; ERRG2; ERRgamma; ESRRG; Estrogen receptor-related protein 3; estrogen related receptor gamma; estrogen-related receptor 3; estrogen-related receptor gamma; FLJ16023; Kiaa0832; mKIAA0832; NR3B3; Nuclear receptor subfamily 3 group B member 3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 2104, 26381, 360896 | |
| -20°C | |
| Lyophilized |
| ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| P62508, P62509, P62510 | |
| Esrrg | |
| E.coli-derived human ESRRG recombinant protein (Position: Q302-V458). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.