Learn More
Estradiol 3-sulfamate, MedChemExpress
Product Code. 16423159
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
Unit Size:
10mg
1mg
20mg
5mg
Product Code. 16423159
Brand: Medchem Express HYU00112
MedChemExpress Estradiol 3-sulfamate (BLE 00084; E2MATE; ES-J 995) is a potent, long-acting, and orally active steroid sulfatase inhibitor; inhibits estrone sulfatase with an IC50 of 251 nM and a Ki of 133 nM.
Estradiol 3-sulfamate (BLE 00084
Specifications
Chemical Name or Material Estradiol 3-sulfamate
CAS 172377-52-5
Color White
Purity Grade Notes Research
Molecular Formula C18H25NO4S
Quantity 1 mg
Synonym BLE 00084 E2MATE ES-J 995
Solubility Information DMSO : 62.5 mg/mL (177.83 mM; Need ultrasonic)
SMILES C[C@@]1([C@H]2O)[C@](CC2)([H])[C@@](CCC3=C4C=CC(OS(N)(=O)=O)=C3)([H])[C@]4([H])CC1
Molecular Weight (g/mol) 351.46
Formula Weight 351.46
Percent Purity 99.5%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less
Research purposes only