Learn More
Estradiol EP Impurity C, Mikromol™
Description
Estradiol EP Impurity C
Specifications
Specifications
| Chemical Name or Material | Estra-1,3,5(10)-triene-3,17beta-diyl Dibenzoate (Estradiol 3,17-Dibenzoate) |
| CAS | 4147-13-1 |
| Analyte or Component Names | Estra-1,3,5(10)-triene-3,17beta-diyl Dibenzoate (Estradiol 3,17-Dibenzoate) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C32 H32 O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Estra-1,3,5(10)-triene-3,17β-diyl Dibenzoate (Estradiol 3,17-Dibenzoate), Estradiol Benzoate Imp. C (EP), Estra-1,3,5(10)-triene-3,17beta-diyl Dibenzoate, Estradiol 3,17-Dibenzoate, Estradiol Benzoate Impurity C |
| InChI Formula | InChI=1S/C32H32O4/c1-32-19-18-26-25-15-13-24(35-30(33)21-8-4-2-5-9-21)20-23(25)12-14-27(26)28(32)16-17-29(32)36-31(34)22-10-6-3-7-11-22/h2-11,13,15,20,26-29H,12,14,16-19H2,1H3/t26-,27-,28+,29+,32+/m1/s1 |
| SMILES | C[C@]12CC[C@H]3[C@@H](CCc4cc(OC(=O)c5ccccc5)ccc34)[C@@H]1CC[C@@H]2OC(=O)c6ccccc6 |
| IUPAC Name | [(8R,9S,13S,14S,17S)-17-benzoyloxy-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-3-yl] benzoate |
| Show More |