missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ESX1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568761
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Mouse: 100%.
Homeobox proteins are transcription factors that contain a helix-turn-helix DNA binding domain termed the homeodomain. ESX1 is an X-linked homeobox gene primarily expressed in the placenta and testis and contains two functional domains: the homeodomain and the proline-rich domain. During embryogenesis, ESX1 is expressed in the extraembryonic tissues, including the endoderm of the visceral yolk sac, the ectoderm of the chorion and the labyrinthine trophoblast of the chorioallantoic placenta. ESX1 can act like a transcriptional repressor to the human oncogene K-ras and treatment of human cancer cells with an ESX1 protein fragment containing the homeodomain reduces the tumorgenicity of cells containing oncogenic K-ras mutations, suggesting ESX1 may be useful as a therapeutic treatment for these cancers.
Specifications
| ESX1 | |
| Polyclonal | |
| Unconjugated | |
| Esx1 | |
| ESX homeobox 1; ESX1; ESX1L; ESX1R; ESX1-related protein; ESXR1; extraembryonic spermatogenesis homeobox 1; extraembryonic, spermatogenesis, homeobox 1; extraembryonic, spermatogenesis, homeobox 1 homolog; homeobox protein ESX1; Homeobox protein ESX1-C; Homeobox protein ESX1-N; RP3-513M9.1; Spx1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 80712 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8N693 | |
| Esx1 | |
| synthetic peptide directed towards the N terminal region of mouse ESX1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction