Learn More
ESYT2 Mouse, anti-Human, Clone: 1B12G2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68374-1-IG-150UL
This item is not returnable.
View return policy
Description
The ESYT2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ESYT2. This antibody recognizes human antigen. The ESYT2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| ESYT2 | |
| 1B12G2 | |
| CHR2SYT, E Syt2, ESYT2, Extended synaptotagmin 2, FAM62B, KIAA1228 | |
| 150 μL | |
| 57488 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG1 |