Learn More
Eszopiclone, Mikromol™
Description
Eszopiclone
Specifications
Specifications
| Chemical Name or Material | Eszopiclone |
| CAS | 138729-47-2 |
| Analyte or Component Names | Eszopiclone |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H17 Cl N6 O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 4-methyl-1-Piperazinecarboxylic acid (5S)-6-(5-chloro-2-pyridinyl)-6,7-dihydro-7-oxo-5H-pyrrolo[3,4-b]pyrazin-5-yl ester, 1-Piperazinecarboxylic acid, 4-methyl-, 6-(5-chloro-2-pyridinyl)-6,7-dihydro-7-oxo-5H-pyrrolo[3,4-b]pyrazin-5-yl ester, (S)-, 5H-Pyrrolo[3,4-b]pyrazine, 1-piperazinecarboxylic acid deriv., (+)-Eszopiclone, (+)-Zopiclone, (S)-Zopiclone, Esopiclone, Estorra, Eszopiclone, Lunesta |
| InChI Formula | InChI=1S/C17H17ClN6O3/c1-22-6-8-23(9-7-22)17(26)27-16-14-13(19-4-5-20-14)15(25)24(16)12-3-2-11(18)10-21-12/h2-5,10,16H,6-9H2,1H3/t16-/m0/s1 |
| SMILES | CN1CCN(CC1)C(=O)O[C@@H]2N(C(=O)c3nccnc23)c4ccc(Cl)cn4 |
| IUPAC Name | [(7S)-6-(5-chloropyridin-2-yl)-5-oxo-7H-pyrrolo[3,4-b]pyrazin-7-yl] 4-methylpiperazine-1-carboxylate |
| Show More |