Learn More
Thermo Scientific Chemicals Eszopiclone
Product Code. 17241316 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17241316 250 mg 250mg
17231316 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17241316 Supplier Thermo Scientific Chemicals Supplier No. 467022500
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 138729-47-2
Molecular Formula C17H17ClN6O3
Specifications
Quantity 250 mg
Solubility Solubility aq. soln.: insoluble.
Melting Point 202.0°C to 206.0°C
Formula Weight 388.81
InChI Key GBBSUAFBMRNDJC-INIZCTEOSA-N
SMILES CN1CCN(CC1)C(=O)O[C@@H]1N(C(=O)C2=NC=CN=C12)C1=CC=C(Cl)C=N1
IUPAC Name (5S)-6-(5-chloropyridin-2-yl)-7-oxo-5H,6H,7H-pyrrolo[3,4-b]pyrazin-5-yl 4-methylpiperazine-1-carboxylate
Molecular Weight (g/mol) 388.81
Chemical Name or Material Eszopiclone