Learn More
Thermo Scientific Chemicals Eszopiclone
Product Code. p-7214813 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 17231316
Brand: Thermo Scientific Chemicals 467020010
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 138729-47-2
Molecular Formula C17H17ClN6O3
Specifications
Quantity 1 g
Solubility Solubility aq. soln.: insoluble.
Melting Point 202.0°C to 206.0°C
Formula Weight 388.81
InChI Key GBBSUAFBMRNDJC-INIZCTEOSA-N
SMILES CN1CCN(CC1)C(=O)O[C@@H]1N(C(=O)C2=NC=CN=C12)C1=CC=C(Cl)C=N1
IUPAC Name (5S)-6-(5-chloropyridin-2-yl)-7-oxo-5H,6H,7H-pyrrolo[3,4-b]pyrazin-5-yl 4-methylpiperazine-1-carboxylate
Molecular Weight (g/mol) 388.81
Chemical Name or Material Eszopiclone