missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ETAA1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557128
This item is not returnable.
View return policy
Description
Immunogen sequence: DMPELFPSKT AHVTDQKEIC TFNSKTVKNT SRANTSPDAR LGDSKVLQDL SSKTYDRELI DAEYRFSPNS NKSNK Highest antigen sequence identity to the following orthologs: Mouse - 45%, Rat - 50%.
ETAA1 (Ewing's tumor-associated antigen 1), also known as ETAA16, is a 926 amino acid cytoplasmic protein that is highly expressed in kidney, brain, liver and Ewing tumor cell lines. ETAA1 undergoes post-translational phosphorylation following DNA damage, most likely by either ATM or ATR, and is suggested to function as a tumour-specific cell surface antigen in Ewing's family of tumour cell lines. The gene encoding ETAA1 maps to human chromosome 2, which consists of 237 million bases, encodes over 1,400 genes and makes up approximately 8% of the human genome. A number of genetic diseases are linked to genes on chromosome 2 including Harlequin icthyosis, sitosterolemia and Alstrom syndrome.
Specifications
| ETAA1 | |
| Polyclonal | |
| Unconjugated | |
| ETAA1 | |
| 5730466H23Rik; AA672646; Etaa1; ETAA16; Ewing tumor associated antigen 1; Ewing tumor-associated antigen 1; ewing's tumor-associated antigen 1; Ewing's tumor-associated antigen 1 homolog; ewing's tumor-associated antigen 16; RGD1560957 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 54465 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NY74 | |
| ETAA1 | |
| Recombinant protein corresponding to Human ETAA1. Recombinant protein control fragment (Product #RP-96111). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction