missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ETEA Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 2 publications
Marca: Novus Biologicals NB100-1296
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
ETEA Polyclonal specifically detects ETEA in Human, Mouse samples. It is validated for Western Blot, Peptide ELISA.
Specifica
| ETEA | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| B17.2L, ETEAFAS-associated factor 2, expressed in T-cells and eosinophils in atopic dermatitis, Fas associated factor family member 2, KIAA0887UBX domain protein 3B, MMTN, NDUNDUFA12L, UBX domain containing 8, UBX domain-containing protein 3B, UBX domain-containing protein 8, UBXD8, UBXN3BProtein ETEA | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 23197 | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:16000 | |
| NP_055428. | |
| FAF2 | |
| Peptide with sequence C-HTEVLFVQDLTDE corresponding to C-Terminus according to NP_055428.1. | |
| 0.1 mg | |
| Primary | |
| Expected from sequence similarity: Mouse, Rat, Dog and Cow. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto