missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ETEA Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA584703

Product Code. 16341655

  • € 578.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: PEPSPDDPES VKIIFKLPND SRVERRFHFS QSLTVIHDFL FSLKESPEKF QIEANFPRRV LPCIPSEEWP NPPTLQEAGL SHTEV Highest antigen sequence indentity to the following orthologs: Rat - 98%, Mouse - 99%.

The protein encoded by this gene is highly expressed in peripheral blood of patients with atopic dermatitis (AD), compared to normal individuals. It may play a role in regulating the resistance to apoptosis that is observed in T cells and eosinophils of AD patients.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ETEA
Polyclonal
Unconjugated
FAF2
2210404d11rik; AI462440; ETEA; expressed in T-cells and eosinophils in atopic dermatitis; FAF2; Fas associated factor family member 2; FAS-associated factor 2; Kiaa0887; mKIAA0887; Protein ETEA; protein expressed in T-cells and eosinophils in atopic dermatitis; RGD1306577; UBX domain containing 8; UBX domain protein 3B; UBX domain-containing protein 3B; UBX domain-containing protein 8; UBX8; UBXD8; UBXN3B; Unknown (protein for MGC:129072)
Rabbit
Antigen affinity chromatography
RUO
23197
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot, Immunocytochemistry, Western Blot
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q96CS3
FAF2
Recombinant protein corresponding to Human FAF2. Recombinant protein control fragment (Product #RP-105374).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.