missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ETFA Antibody (OTI5C4), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70693APC
This item is not returnable.
View return policy
Description
ETFA Monoclonal antibody specifically detects ETFA in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| ETFA | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| OTI5C4 | |
| Western Blot | |
| alpha-ETF, electron transfer flavoprotein alpha-subunit, Electron transfer flavoprotein subunit alpha, mitochondrial, electron-transfer-flavoprotein, alpha polypeptide, EMA, GA2, glutaric aciduria II, MADD | |
| Full length human recombinant protein of human ETFA (NP_000117) produced in E.coli. | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 2108 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction