Learn More
ETFA Monoclonal Antibody (OTI5C4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808409S
This item is not returnable.
View return policy
Description
ETFA Monoclonal antibody specifically detects ETFA in Human samples. It is validated for Western BlotSpecifications
| ETFA | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q5XKR9 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 90736 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI5C4 | |
| Unconjugated | |
| FAM104B | |
| 2010200I21Rik; Alpha-ETF; D9Ertd394e; electron transfer flavoprotein alpha subunit; electron transfer flavoprotein alpha-subunit; electron transfer flavoprotein subunit alpha, mitochondrial; electron transfer flavoprotein, alpha polypeptide; electron transferring flavoprotein, alpha polypeptide; electron-transfer-flavoprotein alpha polypeptide; electron-transfer-flavoprotein, alpha polypeptide; EMA; ETF; ETFA; GA2; glutaric aciduria II; MADD | |
| Full length human recombinant protein of human FAM104B produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |