missing translation for 'onlineSavingsMsg'
Learn More

Ethamsylate, TRC

Product Code. 30009805
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009805 25 mg 25mg
30008399 100 mg 100mg
17982485 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009805 Supplier Toronto Research Chemicals Supplier No. TRCE88995025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2624-44-4
Molecular Formula C6 H6 O5 S . C4 H11 N
Molecular Weight (g/mol) 263.31
Synonym Etamsylate, N-Ethylethanamine 2,5-dihydroxybenzenesulfonate, Benzenesulfonic acid, 2,5-dihydroxy-, compd. with N-ethylethanamine (1:1), Benzenesulfonic acid, 2,5-dihydroxy-, compd. with Et2NH (7CI), Benzenesulfonic acid, 2,5-dihydroxy-, compd. with diethylamine (1:1) (8CI), Diethylamine, 2,5-dihydroxybenzenesulfonate (8CI), Ethanamine, N-ethyl-, 2,5-dihydroxybenzenesulfonate (9CI), 141 MD, 2,5-Dihydroxybenzenesulfonic acid diethylamine salt, Aglumin, Altodor, Biosinon, Cyclonamine, Dicinone, Dicynene, Dicynone, Diethylammonium 2,5-dihydroxybenzenesulfonate, E 141, Eselin, Ethamsylate, Hemsyl, K-Stat, MD 141, Sylate
IUPAC Name 2,5-dihydroxybenzenesulfonic acid;N-ethylethanamine
SMILES CCNCC.Oc1ccc(O)c(c1)S(=O)(=O)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C6H6O5S.C4H11N/c7-4-1-2-5(8)6(3-4)12(9,10)11;1-3-5-4-2/h1-3,7-8H,(H,9,10,11);5H,3-4H2,1-2H3
Formula Weight 263.0827
Percent Purity >95%
Chemical Name or Material Etamsylate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.