missing translation for 'onlineSavingsMsg'
Learn More

Ethanethiol, 97%, Thermo Scientific Chemicals

Catalog Number p-7049775
Click to view available options
Quantity:
25 mL
100 mL
500 mL
This item is not returnable. View return policy
75-08-1
C2H6S
62.13
MFCD00004887
DNJIEGIFACGWOD-UHFFFAOYSA-N
ethyl mercaptan, mercaptoethane, thioethanol, ethyl hydrosulfide, ethyl sulfhydrate, ethyl thioalcohol, thioethyl alcohol, 1-mercaptoethane, aethanethiol, aethylmercaptan
6343
ethanethiol
CCS
This item is not returnable. View return policy
75-08-1
C2H6S
62.13
MFCD00004887
DNJIEGIFACGWOD-UHFFFAOYSA-N
ethyl mercaptan, mercaptoethane, thioethanol, ethyl hydrosulfide, ethyl sulfhydrate, ethyl thioalcohol, thioethyl alcohol, 1-mercaptoethane, aethanethiol, aethylmercaptan
6343
ethanethiol
CCS

Ethanethiol acts as a stabilizer for adhesives and an odorant for natural gas. It is used in the odorless liquefied petroleum gas (LPG) in order to find out the gas leaks. It is used as key starting material for the preparation of ethylsulfonic acid, diethyldisulfide and sodium ethylmercaptide. Further, it is also used in the underground mining industry as stench gas. In organic synthesis, it gives powerful nucleophile set- in the presence of sodium hydroxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -148°C
Color Colorless
Density 0.84 g/mL
Boiling Point 35°C
Flash Point -48°C (-55°F)
Odor Unpleasant
Linear Formula CH3CH2SH
Refractive Index 1.431
Quantity 25 mL
UN Number UN2363
Beilstein 773638
Merck Index 14,3726
Solubility Information Soluble in alcohol,ether,waterMiscible with alcohol,petroleum naphtha,acetone,dilute alkali and ether; Slightly miscible with water.
Formula Weight 62.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethanethiol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'