Learn More
Ethanol, Absolute (200 Proof), Molecular Biology Grade, Fisher BioReagents™
Meets ACS Specifications
Brand: Fisher BioReagents BP2818-500
View more versions of this product
Description
Meets ACS Specifications
- Meets ACS Specifications
- For nucleic acid purification and precipitation
- Ultrapure molecular biology grade ethanol is used for the purification and precipitation of biomolecules such as nucleic acids and proteins
- It is also used in histology to prepare staining and destaining reagents and for dehydrating tissues prior to embedding
Safety and Handling
- Ethanol
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash face, hands and any thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P337+P313-If eye irritation persists: Get medical advice/attention.