missing translation for 'onlineSavingsMsg'
Learn More

ETHE1 Antibody, Novus Biologicals™

Product Code. 18622529 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18622529 100 μg 100µL
18653198 50 μg 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18622529 Supplier Novus Biologicals Supplier No. NBP300355100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ETHE1 Polyclonal antibody specifically detects ETHE1 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ETHE1
Applications Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 1-4 uL/mg of lysate, Immunohistochemistry-Paraffin 1:2500-1:10000
Formulation 0.2 um filtered solution in PBS
Gene Alias D83198, EC 3.1.2.6, ethylmalonic encephalopathy 1, Ethylmalonic encephalopathy protein 1, Hepatoma subtracted clone one protein, HSCOEC 3.-, protein ETHE1, mitochondrial, YF13H12
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human ETHE1 (Accession#: O95571; Leu13-Ala254)
Purification Method Antigen and protein A Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 23474
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.