missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ETHE1 Monoclonal Antibody (GT1621)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527829
This item is not returnable.
View return policy
Description
Positive Control: Neuro2A, GL261, C8D30, NIH-3T3, Raw264.7, C2C1, PC-12, Rat2 Predicted Reactivity: Bovine (90%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ETHE1 is a sulfur dioxygenase that localizes within the mitochondrial matrix. The enzyme functions in sulfide catabolism. Mutations in its gene result in ethylmalonic encephalopathy.
Specifications
| ETHE1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| O95571, Q9DCM0 | |
| Ethe1 | |
| Full length human ETHE1 Recombinant protein. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
| Western Blot, Immunocytochemistry | |
| GT1621 | |
| Unconjugated | |
| Ethe1 | |
| 0610025L15Rik; ETHE1; ETHE1, persulfide dioxygenase; ethylmalonic encephalopathy 1; Ethylmalonic encephalopathy protein 1; Ethylmalonic encephalopathy protein 1 homolog; hepatoma subtracted clone one protein; HSCO; Persulfide dioxygenase ETHE1, mitochondrial; protein ETHE1, mitochondrial; sulfur dioxygenase ETHE1; YF13H12 | |
| Mouse | |
| Protein G | |
| RUO | |
| 23474, 292710, 66071 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction