missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ETHE1 Recombinant Rabbit Monoclonal Antibody (081)

ETHE1 Recombinant Rabbit Monoclonal Antibody (81)
Brand: Invitrogen MA530393
This item is not returnable.
View return policy
Description
Specifications
| ETHE1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| O95571 | |
| Ethe1 | |
| Recombinant Human ETHE1/HSCO Protein (Leu13-Ala254). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA | |
| 081 | |
| Unconjugated | |
| Ethe1 | |
| 0610025L15Rik; ETHE1; ETHE1, persulfide dioxygenase; ethylmalonic encephalopathy 1; Ethylmalonic encephalopathy protein 1; Ethylmalonic encephalopathy protein 1 homolog; hepatoma subtracted clone one protein; HSCO; Persulfide dioxygenase ETHE1, mitochondrial; protein ETHE1, mitochondrial; sulfur dioxygenase ETHE1; YF13H12 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 23474 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction