missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethenoadenosine Antibody (1G4), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB600-442PECY55
This item is not returnable.
View return policy
Description
Ethenoadenosine Monoclonal specifically detects Ethenoadenosine in All Species samples. It is validated for Flow Cytometry.
Specifications
| Ethenoadenosine | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS with 0.05% Sodium Azide | |
| Mouse | |
| Protein G purified | |
| Primary | |
| All species | |
| IgG2 λ |
| Flow Cytometry | |
| 1G4 | |
| Flow Cytometry | |
| C12H13N5O4, N6-Ethenoadenosine | |
| Ethenoadenosine | |
| 0.1 mL | |
| This is specific for ethenoadenosine and ethenodadenosine. | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction